COMH042391EC Combi-Blocks, Inc.

2-Amino-n-benzyl-4-phenylthiophene-3-carboxamide

  • MDL number MFCD03944406
  • Cas Number : 588692-46-0
  • Molecular Weight : 308.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8610
Availability details for
Quantity
  • Availability
  • Add to Cart