COMH042391FA Combi-Blocks, Inc.

[2-(Isopropylthio)ethyl]amine hydrochloride

  • MDL number MFCD18205887
  • Cas Number : 927-69-5
  • Molecular Weight : 155.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8624
Availability details for
Quantity
  • Availability
  • Add to Cart