COMH04239226 Combi-Blocks, Inc.

Methyl n-(4-methylphenyl)-n-(methylsulfonyl)glycinate

  • MDL number MFCD01030685
  • Cas Number : 591731-33-8
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8670
Availability details for
Quantity
  • Availability
  • Add to Cart