COMH0423924B Combi-Blocks, Inc.

1-(1,1-Dioxidotetrahydro-3-thienyl)-3-(4-methoxyphenyl)-1h-pyrazole-5-carboxylic acid

  • MDL number MFCD30188499
  • Cas Number : 2109155-94-2
  • Molecular Weight : 336.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8710

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8710
Availability details for
Quantity
  • Availability
  • Add to Cart