COMH04239273 Combi-Blocks, Inc.

N-(3-Nitrodibenzo[b,d]furan-2-yl)acetamide

  • MDL number MFCD12026779
  • Cas Number : 857785-14-9
  • Molecular Weight : 270.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8750
Availability details for
Quantity
  • Availability
  • Add to Cart