COMH04239276 Combi-Blocks, Inc.

1-Amino-3-(2,6-dimethylphenoxy)propan-2-ol hydrochloride

  • MDL number MFCD29035029
  • Cas Number : 17970-63-7
  • Molecular Weight : 231.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8753
Availability details for
Quantity
  • Availability
  • Add to Cart