COMH042392C7 Combi-Blocks, Inc.

(4-([4-(2-Methoxyphenyl)piperazin-1-yl]sulfonyl)phenyl)amine

  • MDL number MFCD07397893
  • Cas Number : 71454-13-2
  • Molecular Weight : 347.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8837
Availability details for
Quantity
  • Availability
  • Add to Cart