COMH042392D6 Combi-Blocks, Inc.

5-(4-Nitrophenoxymethyl)furan-2-carbaldehyde

  • MDL number MFCD02090891
  • Cas Number : 438221-55-7
  • Molecular Weight : 247.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8853
Availability details for
Quantity
  • Availability
  • Add to Cart