COMH042392F5 Combi-Blocks, Inc.

4,5-Dibromo-3-methylcyclohexane-1,2-dicarboxylic acid

  • MDL number MFCD12197722
  • Cas Number : 65941-85-7
  • Molecular Weight : 344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8884
Availability details for
Quantity
  • Availability
  • Add to Cart