COMH042392F6 Combi-Blocks, Inc.

1-[(3-Chloro-6-nitro-1-benzothien-2-yl)carbonyl]piperazine

  • MDL number MFCD03422692
  • Cas Number : 587852-86-6
  • Molecular Weight : 325.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8885

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8885
Availability details for
Quantity
  • Availability
  • Add to Cart