COMH0423931F Combi-Blocks, Inc.

Methyl 4-(isoxazol-5-ylmethoxy)benzoate

  • MDL number MFCD07757614
  • Cas Number : 890091-84-6
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8926

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8926
Availability details for
Quantity
  • Availability
  • Add to Cart