COMH04239354 Combi-Blocks, Inc.

[2-(Thiomorpholin-4-ylcarbonyl)phenyl]amine

  • MDL number MFCD01569824
  • Cas Number : 664303-98-4
  • Molecular Weight : 222.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8981

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8981
Availability details for
Quantity
  • Availability
  • Add to Cart