COMH04239370 Combi-Blocks, Inc.

2-Methoxy-5-nitrophenyl isothiocyanate

  • MDL number MFCD00060634
  • Cas Number : 71793-51-6
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9009
Availability details for
Quantity
  • Availability
  • Add to Cart