COMH04239393 Combi-Blocks, Inc.

Propyl 5-amino-2-morpholin-4-ylbenzoate

  • MDL number MFCD06752398
  • Cas Number : 876892-54-5
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9044
Availability details for
Quantity
  • Availability
  • Add to Cart