COMH04239397 Combi-Blocks, Inc.

Methyl (4-[(cyclopropylamino)sulfonyl]phenoxy)acetate

  • MDL number MFCD05855284
  • Cas Number : 696632-85-6
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9048
Availability details for
Quantity
  • Availability
  • Add to Cart