COMH042393A9 Combi-Blocks, Inc.

N-(4-Acetyl-2-nitrophenyl)-beta-alanine

  • MDL number MFCD17267863
  • Cas Number : 4662-63-9
  • Molecular Weight : 252.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9067
Availability details for
Quantity
  • Availability
  • Add to Cart