COMH042393CD Combi-Blocks, Inc.

(2-Methyl-4-quinolyl)methanamine hydrochloride

  • MDL number MFCD08061336
  • Cas Number : 1195901-42-8
  • Molecular Weight : 208.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9105
Availability details for
Quantity
  • Availability
  • Add to Cart