COMH042393E5 Combi-Blocks, Inc.

Ethyl 5-(2-furyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD04967320
  • Cas Number : 667920-77-6
  • Molecular Weight : 329.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9130
Availability details for
Quantity
  • Availability
  • Add to Cart