COMH042393E7 Combi-Blocks, Inc.

N-(3-[(Dimethylamino)methylene]-4-oxocyclohexyl)acetamide

  • MDL number MFCD26792687
  • Cas Number : 73983-59-2
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9132
Availability details for
Quantity
  • Availability
  • Add to Cart