COMH042393FB Combi-Blocks, Inc.

3-[(3,5-Dimethylisoxazol-4-yl)methyl]-4-methoxybenzaldehyde

  • MDL number MFCD06801637
  • Cas Number : 887679-08-5
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9149
Availability details for
Quantity
  • Availability
  • Add to Cart