COMH042393FC Combi-Blocks, Inc.

1-[(3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]piperidine-3-carboxylic acid

  • MDL number MFCD12027241
  • Cas Number : 929696-36-6
  • Molecular Weight : 340.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9150
Availability details for
Quantity
  • Availability
  • Add to Cart