COMH04239422 Combi-Blocks, Inc.

5-Bromo-2-[(2-chloro-6-fluorobenzyl)oxy]benzoic acid

  • MDL number MFCD09708239
  • Cas Number : 938257-63-7
  • Molecular Weight : 359.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9188
Availability details for
Quantity
  • Availability
  • Add to Cart