COMH0423942F Combi-Blocks, Inc.

Methyl (4-isothiocyanatophenyl)acetate

  • MDL number MFCD03759881
  • Cas Number : 77055-43-7
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9201
Availability details for
Quantity
  • Availability
  • Add to Cart