COMH04239443 Combi-Blocks, Inc.

(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methylamine

  • MDL number MFCD08691632
  • Cas Number : 915925-36-9
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9221

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9221
Availability details for
Quantity
  • Availability
  • Add to Cart