COMH04239467 Combi-Blocks, Inc.

1-(2-Chloro-6-fluorobenzyl)-5-methyl-1h-pyrazol-3-amine

  • MDL number MFCD04969987
  • Cas Number : 925200-31-3
  • Molecular Weight : 239.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9257
Availability details for
Quantity
  • Availability
  • Add to Cart