COMH04239496 Combi-Blocks, Inc.

6-Methyl-2-(4-propoxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD03074284
  • Cas Number : 438217-48-2
  • Molecular Weight : 321.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9306

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9306
Availability details for
Quantity
  • Availability
  • Add to Cart