COMH042394E8 Combi-Blocks, Inc.

2-[(Phenylsulfonyl)amino]-1,3-benzothiazole-6-carboxylic acid

  • MDL number MFCD09847077
  • Cas Number : 929860-30-0
  • Molecular Weight : 334.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9390
Availability details for
Quantity
  • Availability
  • Add to Cart