COMH0423950F Combi-Blocks, Inc.

2-[(2-Methylbenzyl)thio]acetohydrazide

  • MDL number MFCD04057779
  • Cas Number : 669705-03-7
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9429
Availability details for
Quantity
  • Availability
  • Add to Cart