COMH04239516 Combi-Blocks, Inc.

2-Amino-n-(4-methoxyphenyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD03422611
  • Cas Number : 725226-67-5
  • Molecular Weight : 316.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9436
Availability details for
Quantity
  • Availability
  • Add to Cart