COMH04239525 Combi-Blocks, Inc.

1-(2-Fluorophenyl)-3-methyl-4,5-dihydro-1h-pyrazol-5-one

  • MDL number MFCD00466110
  • Cas Number : 866472-46-0
  • Molecular Weight : 192.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9451
Availability details for
Quantity
  • Availability
  • Add to Cart