COMH04239531 Combi-Blocks, Inc.

4-[(2-Chloro-4-fluorobenzyl)oxy]-3-ethoxybenzaldehyde

  • MDL number MFCD03422472
  • Cas Number : 588713-64-8
  • Molecular Weight : 308.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9463
Availability details for
Quantity
  • Availability
  • Add to Cart