COMH0423954D Combi-Blocks, Inc.

1-(4-Methoxyphenoxy)acetone

  • MDL number MFCD03855315
  • Cas Number : 6698-71-1
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9491
Availability details for
Quantity
  • Availability
  • Add to Cart