COMH04239550 Combi-Blocks, Inc.

4-Allyl-5-(4,5-dimethylthien-3-yl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD03423433
  • Cas Number : 522597-54-2
  • Molecular Weight : 251.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9494
Availability details for
Quantity
  • Availability
  • Add to Cart