COMH04239556 Combi-Blocks, Inc.

2-Bromo-N-(2-methoxy-5-methylphenyl)acetamide

  • MDL number MFCD02974428
  • Cas Number : 130965-93-4
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9500
Availability details for
Quantity
  • Availability
  • Add to Cart