COMH0423958E Combi-Blocks, Inc.

5-(1H-Indol-3-yl)isoxazole-3-carboxylic acid

  • MDL number MFCD00450882
  • Cas Number : 67766-85-2
  • Molecular Weight : 228.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9556
Availability details for
Quantity
  • Availability
  • Add to Cart