COMH042395B1 Combi-Blocks, Inc.

1H-Imidazole-4,5-dicarbohydrazide

  • MDL number MFCD00723482
  • Cas Number : 5423-20-1
  • Molecular Weight : 184.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9592

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9592
Availability details for
Quantity
  • Availability
  • Add to Cart