COMH042395B2 Combi-Blocks, Inc.

N-(3-Chloro-4-fluorophenyl)-2-hydroxybenzamide

  • MDL number MFCD00120193
  • Cas Number : 341018-39-1
  • Molecular Weight : 265.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9593
Availability details for
Quantity
  • Availability
  • Add to Cart