COMH0423964B Combi-Blocks, Inc.

2-Amino-9-[4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-1,9-dihydro-6h-purin-6-one

  • MDL number MFCD00437829
  • Cas Number : 961-07-9
  • Molecular Weight : 267.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9747
Availability details for
Quantity
  • Availability
  • Add to Cart