COMH04239651 Combi-Blocks, Inc.

N-(2,4-Dimethylphenyl)-n-[(4-methylphenyl)sulfonyl]glycine

  • MDL number MFCD02220625
  • Cas Number : 401605-12-7
  • Molecular Weight : 333.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9753
Availability details for
Quantity
  • Availability
  • Add to Cart