COMH04239667 Combi-Blocks, Inc.

(3Z)-1-Phenylquinoline-2,3,4(1h)-trione 3-oxime

  • MDL number MFCD07065507
  • Cas Number : 36412-08-5
  • Molecular Weight : 266.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9775
Availability details for
Quantity
  • Availability
  • Add to Cart