COMH04239678 Combi-Blocks, Inc.

1-(3-Nitrophenyl)-1h-pyrrole

  • MDL number MFCD02665276
  • Cas Number : 4310-42-3
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9792
Availability details for
Quantity
  • Availability
  • Add to Cart