COMH04239692 Combi-Blocks, Inc.

2-Amino-n-(2,5-dimethoxyphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD03017391
  • Cas Number : 440088-43-7
  • Molecular Weight : 332.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9818
Availability details for
Quantity
  • Availability
  • Add to Cart