COMH04239705 Combi-Blocks, Inc.

1,1-Dimethoxy-4-methylpentan-3-one

  • MDL number MFCD18064598
  • Cas Number : 66480-84-0
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-9934
Availability details for
Quantity
  • Availability
  • Add to Cart