COMH042397A4 Combi-Blocks, Inc.

3-(4-Oxo-3,4-dihydro-quinazolin-2-yl)-propionic acid

  • MDL number MFCD03955531
  • Cas Number : 5368-37-6
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2110
Availability details for
Quantity
  • Availability
  • Add to Cart