COMH042397EE Combi-Blocks, Inc.

4-[(2-Oxopyrrolidin-1-yl)methyl]benzoic acid

  • MDL number MFCD06655630
  • Cas Number : 852934-02-2
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4402
Availability details for
Quantity
  • Availability
  • Add to Cart