COMH04239829 Combi-Blocks, Inc.

3-(Phenylthio)propan-1-amine, HCl

  • MDL number MFCD08447166
  • Cas Number : 2015-09-0
  • Molecular Weight : 203.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8417
Availability details for
Quantity
  • Availability
  • Add to Cart