COMH0C9556DF Combi-Blocks, Inc.

3-(1-Hydroxypropyl)benzoic acid

  • MDL number MFCD31653737
  • Cas Number : 1780968-79-7
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6012
Availability details for
Quantity
  • Availability
  • Add to Cart