COMH0C95576E Combi-Blocks, Inc.

4-[1-(Ethoxycarbonyl)cyclopropyl]phenylboronic acid

  • MDL number MFCD30741984
  • Cas Number : 1678539-52-0
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6142
Availability details for
Quantity
  • Availability
  • Add to Cart