COMH0C955771 Combi-Blocks, Inc.

Benzyl N-[2-(methylcarbamoyl)ethyl]carbamate

  • MDL number MFCD30569721
  • Cas Number : 123494-21-3
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4484
Availability details for
Quantity
  • Availability
  • Add to Cart