COMH0C955808 Combi-Blocks, Inc.

2-(Methylsulfanyl)pyridine-3-carbonitrile

  • MDL number MFCD00828729
  • Cas Number : 219766-02-6
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4564
Availability details for
Quantity
  • Availability
  • Add to Cart